Moticam® A Series Cameras, Motic
Supplier: Motic
1100600101412 1100600101292 1100600101302 1100600101443 76538-694 1100600101463 1100600101292 1100600101302 1100600101412 1100600101443
76538-686EA 387.73 CAD
76538-686 76538-688 76538-690 76538-692 76538-694 CA76538-694 CA76538-688 CA76538-690 CA76538-686 CA76538-692
Moticam® A Series Cameras, Motic
Cameras
The Moticam® A series camera is an education microscopy camera line, ranging from 1-16 MP.
  • High-resolution USB cameras
  • Affordable with high-quality imaging
  • All-in-one-box with accessories and software included
  • Multi-platform software- runs on IOS, Android, Windows, Linux
Moticam® microscope cameras come with the accessories needed to make your microscope digital, including the necessary software, USB cable, and a calibration tool. A Moticam can be connected to any microscope through the multiple attachment options available from eyepiece adaptersMore Product Information