Supplier: Cytiva
6756-2502
CA76532-354PK 203 CAD
CA76532-354
Filters Syringe Filters
Whatman puradisc syringe filter with RC membrane is an excellent choice for any application requiring a chemically resistant, hydrophilic, low protein binding membrane. This filter can be used for both aqueous and aggressive organic solvents.
  • Regenerated cellulose membrane exhibits low extractables and protein binding
  • Convenient syringe format streamlines filtration workflow and saves time
  • Rugged heat-welded construction is autoclavable
  • Choice of 13 or 25 mm syringe filter diameter; choice of 0.2 or 0.45 μm pore size
  • Inlet, female Luer lock (FLL); outlet:
More Product Information