CytoFLEX SRT Bench Top Cell Sorter, Beckman Coulter
Supplier: Beckman Coulter
C71883 C71884 C71885 C71886 C71887 C71888 C71889
CA76521-314EA 697231.38 CAD
CA76521-314 CA76521-316 CA76521-318 CA76521-320 CA76521-322 CA76521-324 CA76521-326
CytoFLEX SRT Bench Top Cell Sorter, Beckman Coulter
Cell Counters and Flow Cytometers Flow Cytometers
If you need an approachable instrument for sorting multiple populations to generate high quality material for downstream assays, the CytoFLEX SRT benchtop cell sorter is quick to learn and easy to operate. This extension of the CytoFLEX platform is built on the same optical features as the flow cytometer, but equally important, it continues to adhere to the principles that made multicolor applications accessible to many biomedical research, core research, pharma and biotechs.
  • Exquisite sensitivity for multicolor applications, up to 15 fluorescent colors to
More Product Information