EMnetik Plasmid Purification Kit, Beckman Coulter
Supplier: BECKMAN COULTER CANADA LP CA
C68445
CA76518-678EA 972.1 CAD
CA76518-678
EMnetik Plasmid Purification Kit, Beckman Coulter
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The EMnetik Plasmid Purification Kit is a revolutionary bead-based DNA cleanup kit. Using new SuperSPRI bead technology combined with the microparticle processor EMnetik 24, a user can complete a plasmid prep without having to handle columns.
Benefits of the EMnetik Plasmid Prep System:
- Plasmid recovery of 4 to 7 µg shown for a high copy plasmid.
- No need to handle small columns or use a single-channel pipette.
- Move samples from the lysate to the EMnetik 24 system, and don’t move them again until final elution.
- Intuitive user interface removes guesswork by providing clear, step-by-step instructions.