Membrane Filter Funnels, Microsart® 100/250 Funnels, Sartorius
Supplier: Sartorius
16A0710N 16A0725N
CA76517-846PK 239.99 CAD
CA76517-846 CA76517-848
Membrane Filter Funnels, Microsart® 100/250 Funnels, Sartorius
Funnels Membrane Filter Funnels
PP housing.
  • Reliable results-use a new, sterile funnel for each test for certain avoidances of cross contamination
  • Time saving-kust change the funnel, rather than spending time sanitizing it
  • Simpler handling-no more holding of hot funnels. And you can see when filtration has been completed, particularly useful when using manifolds in routine testing
  • For filter diameter: 47 mm
Microsart® funnels are sterile 100 and 250 ml respectively plastic funnels. They allow quick performance of the filtrations required in the routine testing of water, food andMore Product Information