Microsart® Vacuum Filtration Manifolds, Sartorius
Supplier: Sartorius
168M1MS 168M1SS100 168M1SS500 168M2BS100 168M2MS 168M3BS100 168M3BS250 168M3MS 168M3SS100 168M3SS500 168M6BS100 168M6BS250 168M6MS 168M6SS100 168M6SS500
CA76517-804EA 2615.63 CAD
CA76517-804 CA76517-806 CA76517-808 CA76517-810 CA76517-812 CA76517-814 CA76517-816 CA76517-818 CA76517-820 CA76517-822 CA76517-824 CA76517-826 CA76517-828 CA76517-830 CA76517-832
Microsart® Vacuum Filtration Manifolds, Sartorius
Manifolds
Microsart® Manifolds accurately detect and quantify microorganisms in liquid samples. Simply select the proper manifold for your individual microbiological testing procedure, choose between reusable and single-use funnels or filtration units, and then decide between different sizes of manifolds and enjoy the manifold that adapts to your needs.
  • 100% stainless steel for minimised cleaning effort and full autoclavability without taking apart
  • Sterile venting in critical applications reduces risk of secondary contamination
  • Quick connection adapters for fast
More Product Information