Membrane Filters with Grid, Sartorius
Supplier: Sartorius
1140347ACN 1140347ACR 1140347N 1140447ACN 1140447ACR 1140447N 1140450ACN 1140450N 1140650ACR 1140650N 1140747ACN 1140747ACR 1140747N 1140747R 1140750ACN 1140750ACR 1140750N 114H647ACN 114H647ACR 114H650ACR 1300447ACN 1300447ACR 1300447N 1300450N 1300547ACN 1300547N 1300550ACN 1300550ACR 1300550N 1300647ACN 1300647ACR 1300647N 1300647R 1300650ACN 1300650ACR 1300650N 130H647ACN 130H647ACR 130H650ACR
CA76517-556PK 202.75 CAD
CA76517-556 CA76517-558 CA76517-560 CA76517-564 CA76517-566 CA76517-568 CA76517-570 CA76517-572 CA76517-588 CA76517-590 CA76517-596 CA76517-598 CA76517-600 CA76517-602 CA76517-604 CA76517-606 CA76517-608 CA76517-614 CA76517-616 CA76517-620 CA76517-630 CA76517-632 CA76517-634 CA76517-636 CA76517-642 CA76517-644 CA76517-646 CA76517-648 CA76517-650 CA76517-656 CA76517-658 CA76517-660 CA76517-662 CA76517-664 CA76517-666 CA76517-668 CA76517-674 CA76517-676 CA76517-678
Membrane Filters with Grid, Sartorius
Filters Membrane Filters
Cellulose nitrate hydrophilic, gridded membrane filters for colony counting, particle testing and microscopy, with outstanding recovery rates for microorganisms.
Membranes with 0,45 µm pore size are tested and released according to ISO 7704.
Order Now
SPECIFICATIONS
Environmentally Preferable
Water flow rate 320 ml/min (1 bar) 20 ml/min (1 bar) 130 ml/min (1 bar) 70 ml/min (1 bar) 100 ml/min (1 bar)
Max. temperature 130 °C 130 °C 130 °C 130 °C 130 °C 130 °C