Benchtop Centrifuges, Megafuge ST1 and ST1R Plus and Packages, Thermo Scientific
Supplier: Thermo Fisher Scientific
75009230 75009271 75016002 75016003 75016010 75017002 75017003 75016026US 75009230CAP 75009271CAP 75016010CAP 75016003CAP 75016002CAP 75017003CAP 75017002CAP 75016026USCAP
76470-414EA 13066.92 CAD
76470-414 76470-418 76470-426 76470-428 76470-430 76470-438 76470-442 76470-454 CA76501-574 CA76501-576 CA76501-604 CA76501-610 CA76501-612 CA76501-618 CA76501-620 CA76501-602
Benchtop Centrifuges, Megafuge ST1 and ST1R Plus and Packages, Thermo Scientific
Centrifuges Benchtop Centrifuges
The Thermo Scientific™ Megafuge ST1/ST1R Plus benchtop centrifuges offer intuitive controls and versatility for power at every turn.
  • Auto-Lock rotor system
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switching between applications. Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life. Biocontainment solution seals with a snap; eliminates screw capsMore Product Information