extraktLAB fracTron-1000 Falling Film Evaporator, United Science
Supplier: United Science
10-0080
CA76501-202EA 466264.46 CAD
CA76501-202
extraktLAB fracTron-1000 Falling Film Evaporator, United Science
Distillation Systems Automatic Distillation Systems
Three-stage fractional solvent distillation system for rapid removal of volatiles from liquid mixtures.
  • 3-stage fractional separation
  • Inert carrier gas
  • One year warranty
fracTRON is a three-stage fractional solvent distillation system designed for rapid and complete removal of volatiles from liquids. It supports throughput capacity of 40 gallons per hour in a continuous process flow.
The fracTRON can support three independent fractional separations with independent mass flow and temperature control. An inert carrier gas is incorporated toMore Product Information