KleenGuard™ G10 2PRO™ Nitrile Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
54420 54421 54422 54423 54424
CA76501-076BX 57.22 CAD
CA76501-076 CA76501-078 CA76501-080 CA76501-082 CA76501-084
KleenGuard™ G10 2PRO™ Nitrile Gloves, Kimberly-Clark
Gloves
KleenGuard™ G10 2PRO™ nitrile gloves allow for maximum productivity and safety, for you and your employees.
  • Blue nitrile gloves
  • Powder-free, ambidextrous, and touchscreen compatible
  • Gloves meet U.S. FDA regulations for food contact applications
  • Textured fingertips give employees a grippable surface for dexterity and control while performing sensitive tasks
  • Beaded cuffs prevent roll-down, help alleviate concerns of contamination, and allow for superior donning
  • Gloves are not made with natural rubber latex
KleenGuard™ Personal Protective Equipment (PPE)More Product Information