KleenGuard™ G10 Comfort Plus™ Nitrile Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
54185 54186 54187 54188 54189
CA76501-066BX 48.64 CAD
CA76501-066 CA76501-068 CA76501-070 CA76501-072 CA76501-074
KleenGuard™ G10 Comfort Plus™ Nitrile Gloves, Kimberly-Clark
Gloves
When searching for a glove that is designed for superior strength and tactile sensitivity, KleenGuard™ G10 Comfort Plus™ nitrile gloves are an excellent choice.
  • Light blue nitrile gloves
  • Powder-free, ambidextrous, and touchscreen compatible
  • Comfortable, protective, and user-friendly
  • Advanced formulation delivers superior strength and tactile sensitivity
KleenGuard™ Personal Protective Equipment (PPE) enables rather than inhibits, allowing for maximum productivity and safety for you and your employees. These 9" ambidextrous gloves feature a beaded cuffMore Product Information