Minisart® PES Venting and Gas Filtrations
Supplier: Sartorius
1757GK 1757GQ 1757HK 1757HQ
CA76474-438PK 252.22 CAD
CA76474-438 CA76474-440 CA76474-442 CA76474-444
Minisart® PES Venting and Gas Filtrations
Filters Syringe Filters
Minisart® PES with hydrophobic polyethersulfone (PES) is suitable for venting or filtration of gases and air.
  • Non-sterile filters for assembly
  • High chemical compatibility
  • Suitable as venting filter
Units can be sterilized by gamma irradiation, autoclaving or EO. Non-sterile Minisart® PES- syringe filters are supplied in a pouch in one pack. This reduces waste and makes the handling easier whenever the application allows venting under non-sterile conditions.
In addition the non-sterile filters can be assembled and sterilised by the user. TheMore Product Information