Laboratory Glassware Washers, Model PLW 8617, Miele
Supplier: Miele
62861701USA 62861702USA 62861703USA 62861704USA 62861705USA 62861706USA 62861707USA 62861708USA 99999547 99999548 99999537 99999538 99999539 99999540 99999541
CA76472-500EA 88373.05 CAD
CA76472-500 CA76472-502 CA76472-504 CA76472-506 CA76472-508 CA76472-510 CA76472-512 CA76472-514 CA77445-722 CA77445-724 CA77445-704 CA77445-706 CA77445-708 CA77445-710 CA77445-712
Laboratory Glassware Washers, Model PLW 8617, Miele
Glassware Washers
The PLW 8617 laboratory washer can accommodate 216 laboratory flasks, 588 vials, or 294 pipettes. The large glassware washer is flexible and simple to use, it has modular load carriers and SimpleLoad system. This efficient unit has a single door, 1150 mm (3' 91/4") wide, a usable capacity of 351 litres. PLW 8617 provides reliable results, high pump performance with a variable-speed pump and features spray arm monitoring and a conductivity meter.
  • Innovative 2-pump system for maximum performance
  • New flexible and modular (SimpleLoad)racking system
  • Excellent
More Product Information