CyBio® FeliX Automated Pipet Robots, Analytik Jena US
Supplier: Analytik Jena CA
OL5015-24-100 OL5015-25-500 OL5015-25-505
CA76469-566EA 60487.23 CAD
CA76469-566 CA76469-568 CA76469-570
CyBio® FeliX Automated Pipet Robots, Analytik Jena US
Automated Liquid Handling Systems
Flexible pipetting platform for fully automated single to multichannel liquid handling.
  • Flexible pipettor with 1 to 384 channels with automatic loading of pipetting tools
  • Unique and compact deck design, 12 positions on two levels
  • CHOICE™ technology for pipetting from 500 nl to 1 ml
  • Highest precision and accuracy for various applications in all formats
  • Closed stand-alone system or as an open system for placing in laminar flow hood and integration
The CyBio® FeliX offers maximum performance for the automation of your cellular assays such as mediaMore Product Information