Laser Microdissection (LMD) System, Leica Microsystems
Supplier: Leica Microsystems
11600288 11505158 11505189 11505215 11600312 11505188 11505151 11505190 11505191
CA103501-506PK 417.78 CAD
CA103501-506 CA76414-774 CA76414-776 CA76414-778 CA76414-780 CA76463-316 CA76463-318 CA76463-320 CA76463-322 CA76463-324
Laser Microdissection (LMD) System, Leica Microsystems
Microtomes and Microtome Assemblies Microtomes
We move the laser, not the sample. And we use gravity for collection. LMD systems provides perfectly cut, contamination-free, analysis-ready dissectates.
  • Unique laser-guidance based dissection method to collect dissectates via gravity – contact-free and contamination-free
  • High-precision optics to steer the laser beam along the desired cut line, leaving clean cuts of contamination-free isolates
  • LMD SmartCut objectives made specifically for the LMD system
  • Save on consumables, as the Leica LMD utilizes standard PCR tubes to capture isolated samples. You will
More Product Information