AcroPrep™ 24-Well Filter Plates, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
97026 97026 97016 97027 97017 97029 97030 97031 97032 97033 97034 97035 97036 97047 97048 97049 97050 97051 97052 97053 97054 97055 97056 97057 97058 97059 97060 97061 97062 97029 97030 97031 97032 97033 97034 97035 97036 97047 97048 97049 97050 97051 97052 97053 97054 97055 97056 97057 97058 97059 97060 97061 97062
76360-446EA 870.88 CAD
76360-446 CA76360-446 CA76360-448 CA76360-450 CA76360-452 76457-240 76457-242 76457-244 76457-246 76457-248 76457-250 76457-252 76457-254 76457-256 76457-258 76457-260 76457-262 76457-264 76457-266 76457-268 76457-270 76457-272 76457-274 76457-276 76457-278 76457-280 76457-282 76457-284 76457-286 CA76457-240 CA76457-242 CA76457-244 CA76457-246 CA76457-248 CA76457-250 CA76457-252 CA76457-254 CA76457-256 CA76457-258 CA76457-260 CA76457-262 CA76457-264 CA76457-266 CA76457-268 CA76457-270 CA76457-272 CA76457-274 CA76457-276 CA76457-278 CA76457-280 CA76457-282 CA76457-284 CA76457-286
AcroPrep™ 24-Well Filter Plates, Cytiva (Formerly Pall Lab)
Microplates Filter Plates
The Acroprep 24-well filter plate utilizes Pall’s proprietary high-performance multi-layer filter media and membranes to offer time savings, strong performance claims and streamlined workflow improvements in a 24-well plate format.
  • Intrinsic Acroprep plate and membrane properties minimize sample loss from non-specific binding
  • Dual layer clarification plate contains a depth filter plus 0.65/0.2 μm Supor EKV PES membrane and allow clarification of large cellular debris prior to filtration through the 0.65/0.2 μm EKV Supor membrane
  • Dual layer clarification
More Product Information