Whatman™ Grade GF 10 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Supplier: Cytiva
10370302 10370319 10370305 10370320 10370308 10370302 10370305 10370308 10370319 10370320 10370393 10370391
89187-082EA 1397.35 CAD
89187-082 CA11013-452 CA11013-464 CA11013-542 CA11013-840 CA11013-860 89428-194 89428-196 89428-198 10034-846 10034-848 10034-850 10034-852 10034-854 CA89187-082 CA89428-194 CA89428-196 CA89428-198 CA76333-760 CA76423-724
Whatman™ Grade GF 10 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Filters Glass Fiber Filters
These glass microfibre filters made from borosilicate glass are strengthened with an organic binder to provide high mechanical stability.
  • Medium filtration speed
  • Can be used as a roll filter in automated applications
These filters can be used as a weighing aid during infrared weighing or for automatic sampling applications. They also come in reel format for use as a roll filter in automatic air filtration units.