EasyReach™ Cleaning Tools, Contec®
Supplier: Contec
MEQT0001 PSME0001 MEQT0021 MEQT0022 2685LF 2665LF MEQT0002SZ 2685P
89094-578CS 573.28 CAD
89094-578 89094-580 89094-582 89094-584 89107-994 76417-132 76417-134 CA76417-120 CA76417-122 CA76449-778 76480-378
EasyReach™ Cleaning Tools, Contec®
Mops
The lightweight, stainless steel EasyReach™ Cleaning Tool is designed for cleaning isolators, laminar air flow hoods, glove boxes, and other mini environments. Rectangular shape makes overlapping straight-line action easy to achieve. Hinged joint and lightweight design makes one-handed cleaning comfortable and improves access to hard-to-reach areas.
  • Rounded tool edges and strong, durable, double-layer pads ensure equipment is not damaged during use
  • Lightweight EasyReach™ Tool can easily be manipulated, is comfortable to use, and minimizes fatigue
  • All
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable