Mag-Bind® Viral DNA/RNA 96 Kit, Omega Bio-tek

Supplier: OMEGA BIO-TEK CA

M6246-03
CA76414-002EA 4509.84 CAD
CA76414-002
Mag-Bind® Viral DNA/RNA 96 Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

Mag-Bind® Viral DNA/RNA Kit is designed for the rapid and reliable isolation of viral RNA and viral DNA from whole Blood, serum, plasma, saliva and other body fluids


The Mag-Bind® magnetic beads technology enables purification of high-quality nucleic acids that is free of proteins, nucleases, and other impurities. In addition to easily being adapted with automated systems, this procedure can also be scaled up or down, allowing for the purification from various amounts of starting materials. The purified nucleic acid is ready for direct use in downstream applications, such as amplification or other enzymatic reactions.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Target molecule Copurification of DNA & RNA,DNA,RNA
Refine target molecule Viral RNA,Viral DNA
Sample type Swabs,Blood,Biofluids (serum, plasma, saliva)
Method / Format Magnetic beads
Prep size Scalable
Sample size 50 - 200 μl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR