DMiL Inverted Microscope Packages, Leica Microsystems
Supplier: Leica Microsystems
11541544 11521288 11504254 11504127 11506408 11506272 11506298 11506203 11504190 11504174 11504179 11504208 11504117
CA10037-632EA 809.75 CAD
CA10037-632 CA76377-688 CA76377-690 CA76377-696 CA76377-698 CA76377-700 CA76377-702 CA76377-704 CA76377-706 CA76377-708 CA76377-710 CA76377-712 CA76377-714 CA10037-662
DMiL Inverted Microscope Packages, Leica Microsystems
Microscopes
The Leica DM IL is a fluorescence ready digital imaging solution designed for use in a variety of laboratory settings including cell culture, micromanipulation, immuno staining and routine live imaging.
  • Inverted laboratory microscope with LED illumination
  • High-performance optics
  • Ergonomic design
  • 3W white light LED illumination
The DM IL LED Inverted Microscope Lab Package is a robust inverted microscope with LED illumination designed for years of use without the need to replace bulbs.
The DM IL LED is fully configured for both brightfield andMore Product Information