SD Pro™ Primate Probiotic Tablets, Bio-Serv
Supplier: Bio-Serv
PB100-BLK-B PB100-BLK PB100-BLK-B
CA10798-916PK 213.25 CAD
CA10798-916 76285-294 76285-296 CA76353-786 CA76285-296
SD Pro™ Primate Probiotic Tablets, Bio-Serv
Animal Supplies Animal Feed
SD Pro™ Primate Probiotic Tablets are a non-human primate origin bacteria probiotic featuring Lactobacillus reuteri.
Ingredients: Sucrose, Dextrose, Strawberry Flavor, Yeast Culture, Hydrolyzed Yeast, Calcium Silicate, Magnesium Stearate, Dried Lactobacillus Reuteri, Fermentation Product, Soybean Oil, Sodium Aluminosilicate.
Feeding Directions: <1 kg primate: ¼ tablet daily; 1-2 kg primate: ½ tablet daily; 3-10 kg primate: 1 tablet daily; >10 kg primate: 2-4 tablets daily. Note: If non-human primate is on antibiotic therapy, administer PrimiOtic 4–6 hours after antibiotic dosage.
More Product Information