V Agar, for Gardnerella vaginalis
Supplier: HARDY DIAGNOSTICS CA
A80
CA76339-340PK 199.5 CAD
CA76339-340
V Agar, for Gardnerella vaginalis
Media for Microbiology Prepared Media for Microbiology
(Columbia Base with 5% Human Blood) For the detection of Garnerella vaginalis.
Ordering information: This product has a short shelf life.
Delivery information: This product ships from our California warehouse only.
Caution: Use Biohazard Precautions.