Cubis® II Essential Premium High Capacity Laboratory Balances, without Draft Shield, Sartorius
Supplier: Sartorius
MCE36200S0-S00 MCE36201S-0-MTS MCE20201S0-S00 MCE70201S2S000 MCE50201S2S000 MCE36201S2S000 MCE36201P2S000 MCE20201S2S000 MCE11201S2S000 MCE70200S2S000 MCE36200S2S000
CA76333-816EA 8731.88 CAD
CA76333-816 CA76333-818 CA76333-942 CA76334-402 CA76334-404 CA76334-406 CA76334-408 CA76334-410 CA76334-412 CA76334-414 CA76334-416
Cubis® II Essential Premium High Capacity Laboratory Balances, without Draft Shield, Sartorius
Balances and Scales
These Cubis® II premium high capacity laboratory balances are ideal for high-performance portfolio of both lab weighing hardware and software.
  • Quick and easy, motorized, manual-levelling function, integrated climate sensors, individual sample holders
  • Error-free operation with individual QApp workflows and full end-to-end data integrity
  • Outstanding service support integrated status center, integrated service functions preventive maintenance on accredited standards
  • User Management provides complete access control and configured in accordance with password
More Product Information