Cubis® II Essential Premium Precision Laboratory Balances, Automatic, Standard Version, Sartorius
Supplier: Sartorius
MCE1203S2S00R MCE623S2S00R MCE2203SR-S00 MCE2203P2S00E MCE1203S2S00E MCE623S2S00E MCE623P2S00E MCE323S2S00E MCE5203S2S00I MCE5203P2S00I MCE3203S2S00I MCE2203S2S00I MCE1203SR-S00 MCE3203SR-S00 MCE5203S2S00A MCE5203P2S00A MCE3203S2S00A MCE2203S2S00A MCE2203P2S00A MCE1203S2S00A MCE623S2S00A MCE623P2S00A MCE323S2S00A MCE5203S2S00E MCE2203P2S00I MCE1203S2S00I MCE623S2S00I MCE623P2S00I MCE323S2S00I MCE5203S2S00R MCE5203P2S00R MCE3203S2S00R MCE2203S2S00R MCE623P2S00R MCE323S2S00R MCE5203S2S00U MCE5203P2S00U MCE3203S2S00U MCE2203S2S00U MCE2203P2S00U MCE1203S2S00U MCE623S2S00U MCE623P2S00U MCE323S2S00U MCE5202S2S00A MCE5202S2S00E MCE5202S2S00I MCE5202S2S00R MCE5202S2S00U MCE2203P2S00R MCE2203S2S00E MCE3203S2S00E MCE5203P2S00E
CA76333-824EA 9647.48 CAD
CA76333-824 CA76333-914 CA76333-916 CA76333-946 CA76334-124 CA76334-126 CA76334-128 CA76334-130 CA76334-132 CA76334-134 CA76334-136 CA76334-138 CA76334-140 CA76334-148 CA76334-156 CA76334-316 CA76334-318 CA76334-320 CA76334-322 CA76334-324 CA76334-326 CA76334-328 CA76334-330 CA76334-332 CA76334-334 CA76334-336 CA76334-338 CA76334-340 CA76334-342 CA76334-344 CA76334-346 CA76334-348 CA76334-350 CA76334-352 CA76334-354 CA76334-356 CA76334-358 CA76334-360 CA76334-362 CA76334-364 CA76334-366 CA76334-368 CA76334-370 CA76334-372 CA76334-374 CA76334-418 CA76334-420 CA76334-422 CA76334-424 CA76334-426 CA76377-030 CA76377-040 CA76377-068 CA76377-118
Cubis® II Essential Premium Precision Laboratory Balances, Automatic, Standard Version, Sartorius
Balances and Scales
These MCE series essential user interface balances have a large, high-contrast touch display with factory-installed essential weighing applications for easy operation.
  • Error-free operation with individual QApp workflows, motorized auto-leveling*
  • Full end-to-end data integrity
  • Outstanding service support integrated status center, integrated service functions preventive maintenance on accredited standards
  • User Management provides complete access control and configured in accordance with password policy
  • Gesture control allows user to open and close the
More Product Information