Cubis® II Advanced Premium Precision Laboratory Balances, Automatic, Standard Version, Sartorius
Supplier: Sartorius
MCA2203SR-S00 MCA3203SE-S00 MCA5203SR-S00 MCA5203S-2S00-A MCA5203P-2S00-A MCA3203S-2S00-A MCA2203S-2S00-A MCA2203P-2S00-A MCA1203S-2S00-A MCA623S-2S00-A MCA1203S-2S00-E MCA623S-2S00-E MCA623P-2S00-E MCA323S-2S00-E MCA5203S-2S00-I MCA5203P-2S00-I MCA3203S-2S00-I MCA2203S-2S00-I MCA2203P-2S00-I MCA1203S-2S00-I MCA623S-2S00-I MCA623P-2S00-I MCA323S-2S00-I MCA5203S-2S00-R MCA2203P-2S00-R MCA1203S-2S00-R MCA623S-2S00-R MCA623P-2S00-R MCA323S-2S00-R MCA5203S-2S00-U MCA5203P-2S00-U MCA3203S-2S00-U MCA2203S-2S00-U MCA2203P-2S00-U MCA1203S-2S00-U MCA623S-2S00-U MCA623P-2S00-U MCA323S-2S00-U MCA623SR-S00 MCA623P-2S00-A MCA5203S-2S00-A MCA5203P-2S00-R MCA3203S-2S00-R MCA2203S-2S00-R MCA623SE-S00 MCA1203SE-S00
CA76333-804EA 18857.32 CAD
CA76333-804 CA76333-806 CA76333-810 CA76333-912 CA76333-928 CA76334-018 CA76334-020 CA76334-022 CA76334-024 CA76334-026 CA76334-028 CA76334-030 CA76334-032 CA76334-034 CA76334-036 CA76334-038 CA76334-040 CA76334-042 CA76334-044 CA76334-046 CA76334-048 CA76334-050 CA76334-052 CA76334-054 CA76334-056 CA76334-058 CA76334-060 CA76334-062 CA76334-064 CA76334-066 CA76334-068 CA76334-070 CA76334-072 CA76334-074 CA76334-076 CA76334-078 CA76334-080 CA76334-082 CA76334-084 CA76334-086 CA76334-088 CA76334-090 CA76334-092 CA76334-094 CA76334-142 CA76334-228 CA76334-230 CA76334-232 CA76334-234 CA76334-236 CA76334-238 CA76377-062 CA76377-162 CA76377-164 CA76377-288
Cubis® II Advanced Premium Precision Laboratory Balances, Automatic, Standard Version, Sartorius
Balances and Scales
These MCA series advanced user interface laboratory balances have fully customizable hardware, software, and connectivity including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QApp packages for optional software extension.
  • Quick and easy, motorized, auto-levelling function, integrated climate sensors, individual sample holders
  • Error-free operation with individual QApp workflows and full end-to-end data integrity
  • Outstanding service support integrated status center, integrated service functions
More Product Information