Precision Balances, Entris® II Essential, Sartorius
Supplier: Sartorius
BCE1202-1S BCE1202I-1S BCE2201-1S BCE2201I-1S BCE2202-1S BCE2202I-1S BCE223-1S BCE223I-1S BCE3202-1S BCE3202I-1S BCE323-1S BCE323I-1S BCE4202-1S BCE4202I-1S BCE423-1S BCE423I-1S BCE5201-1S BCE5201I-1S BCE6200-1S BCE6200I-1S BCE6202-1S BCE6202I-1S BCE622-1S BCE622I-1S BCE623-1S BCE623I-1S BCE653-1S BCE653I-1S BCE8200-1S BCE8200I-1S BCE8201-1S BCE8201I-1S BCE822-1S BCE822I-1S
CA76333-828EA 1893.56 CAD
CA76333-828 CA76333-830 CA76333-836 CA76333-838 CA76333-840 CA76333-842 CA76333-844 CA76333-846 CA76334-162 CA76334-164 CA76334-166 CA76334-168 CA76334-170 CA76334-172 CA76334-174 CA76334-176 CA76334-178 CA76334-180 CA76334-182 CA76334-184 CA76334-186 CA76334-188 CA76334-190 CA76334-192 CA76334-194 CA76334-196 CA76334-202 CA76334-204 CA76334-206 CA76334-208 CA76334-210 CA76334-212 CA76334-214 CA76334-216
Precision Balances, Entris® II Essential, Sartorius
Balances and Scales
These Entris® II laboratory balances are ideal for basic weighing tasks.
  • Highly accurate results guaranteed via Monolithic weigh cell technology
  • Fastest stabilization time in its class using state-of-the-art weighing sensors
  • Future-proof USB Type C interface and industry-proof RS232 9-pin interface
  • Prevents cross contamination with wipe clean design and easy-to-remove parts
  • isoCAL (internal calibration and adjustment) for total assurance of accurate weighing results
  • Best repeatability with rectangular weighing pan
  • Overload protection with rugged design
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Voltage 100 - 240 V