Quick-Seal Ultracentrifugation Tubes, Beckman Coulter
Supplier: Beckman Coulter
360970 345830 360974 357343 360973 344338 355643 343665 301112 345831 345775 358980 358125 344320 344622 349621 358120 361707 355616 355636 348176 360972 361659 355646 345777 356957 363962 348177 343777 344323 344093 348184 363961 344075
CA76333-110KT 3003.51 CAD
CA76333-110 CA76333-112 CA76333-114 CA76333-116 CA76333-118 CA76333-120 CA76333-122 CA76333-124 CA76333-126 CA76333-128 CA76333-130 CA76333-132 CA76333-134 CA76333-136 CA76333-232 CA76333-234 CA76333-236 CA76333-238 CA76333-240 CA76333-242 CA76333-312 CA76333-752 CA76333-754 CA76333-756 CA76378-478 CA76378-480 CA76378-482 CA76333-764 CA76333-766 CA76333-768 CA76333-776 CA76333-778 CA76333-780 CA76333-782 CA76333-786
Quick-Seal Ultracentrifugation Tubes, Beckman Coulter
Tubes Centrifuge Tubes
Quick-seal tubes eliminate most of the steps involved in capping tubes and cut handling time in half.
  • Different types of materials
  • Various configurations
  • Wide selection
The top of the tube is either dome-shaped or bell-shaped with a 3 mm-long inlet through which the tube is filled. The filled tube is heat-sealed using the hand-held sealer (required). The highly reliable seal achieved makes these tubes ideal for sample storage and for working with radioactive or pathogenic samples.
Beckman Coulter quick-seal tubes are designed for use in swingingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable