Whatman™ Nuclepore™ Polycarbonate Hydrophilic Membranes
Supplier: Cytiva
10418701 10418704 10418706 10418712 10418718 10418731 10418739 10418806 10418812 10418818 10418318 10417101 10417104 10417106 10417112 10417114 10417118 10417206 10418401 10418406 10418412 10418501 10418506 10418512 10418514 10417212 10417301 10417304 10417306 10417309 10417312 10417401 10417406 10417412 10417414 10417501 10417506 10417512
CA76457-178PK 121.78 CAD
CA76457-178 CA76457-188 CA76457-194 CA76457-196 CA76457-170 CA76457-208 CA76457-172 CA76457-184 CA76457-202 CA76457-206 CA76457-204 CA76332-574 CA76332-642 CA76332-584 CA76332-616 CA76332-628 CA76332-634 CA76332-604 CA76457-174 CA76457-192 CA76457-186 CA76457-190 CA76457-180 CA76457-182 CA76457-210 CA76332-618 CA76332-576 CA76332-644 CA76332-606 CA76332-612 CA76332-620 CA76332-578 CA76332-608 CA76332-622 CA76332-630 CA76332-580 CA76332-610 CA76332-624
Whatman™ Nuclepore™ Polycarbonate Hydrophilic Membranes
Filters Membrane Filters
Nuclepore™ track-etched hydrophilic membrane filters offer a highly defined pore size cut-off in the submicron range, enabling particle deformity measurement and capture of extremely small particles. The membranes have sharply defined pores, high flow rates, and excellent chemical and thermal resistance. They are suitable for a variety of samples and applications, including microscopy, cytology, parasitology, and certain air and oceanography analyses.
  • Track-etched for highly defined pore size and high pore density
  • Treated with polyvinylpyrrolidone (PVP) to
More Product Information