BioClean™ Nitrile Clean Non Sterile RABS/Isolator Gloves, Ansell
Supplier: Ansell Healthcare
CGL20NIT59 CGL10NIT59 CGL36NIT59 CGL10NIT59
76316-253CS 10859.35 CAD
76316-253 76316-255 76316-257 CA76316-255
BioClean™ Nitrile Clean Non Sterile RABS/Isolator Gloves, Ansell
Gloves
Non sterile RABS/Isolator gloves.
  • Tested against ASTM D6978-05 for handling chemo drug
  • 100% water leak tested
  • Ultra-clean surface ensures product protection
  • Autoclave compatible
  • ISO 4 Cleanroom laundered and packaged
  • Resistant to VHP, IPA, and disinfectants
BioClean™ RABS and isolator gloves are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties. Designed for use in injectable manufacturing and pharmaceutical production areas, the CGL series of gloves are cleanroom processed and packed, and are available inMore Product Information
New Product
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Material Nitrile
Finish Non-clean, fully dipped
Cuff style Beaded
Cuff thickness 20 mil
Palm thickness 20 mil