Mycoplasma Agar
Supplier: HARDY DIAGNOSTICS CA
G362
CA76300-102PK 77.09 CAD
CA76300-102
Mycoplasma Agar
Media for Microbiology Prepared Media for Microbiology
For the cultivation of Mycoplasma and the detection of Mycoplasma contamination in biologics and biological processes.
  • Optically clear polystyrene - For distortion free viewing.
  • Heavy-Duty - Reduces breakage during shipping.
  • Stacking rings - Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
Ordering information: Minimum order: 5 Packs
Packaging: Breathable Packaging - Packaged in sleeves of 10 plates in a "breathable" bag that prevents buildup of condensation and excess moisture.