NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina, New England Biolabs
Supplier: New England Biolabs (NEB)
E6420L E6420S
CA76293-214EA 6605 CAD
CA76293-214 CA76293-216
NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina uses a template switching method to generate full length cDNAs directly from single cells or 2 pg to 200 ng RNA, followed by conversion to sequence-ready libraries using the Ultra II FS workflow.
  • Experience unmatched detection of low abundance transcripts
  • Rely on consistent transcript detection for a wide range of input amounts and sample types
  • Observe uniform transcript coverage, regardless of input amount or sample type
  • Use with a variety of RNA inputs, including cultured or primary cells, or total RNA
This unique workflow enables generation of the highest yields from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance.
Ordering information: Please note that adaptors and primers are not included in the kit and are available separately.