Calcium chloride dihydrate ACS

Supplier: INNOVA CHEMICALS CA
Warning

INN890519 INN890520
CA76288-566CS 115 CAD
CA76288-566 CA76288-568
Calcium chloride dihydrate ACS
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
MDL Number: MFCD00149613
CAS Number: 10035-04-8
Merck Index: 13,01660

Order Now

Specification Test Results

Form/Colour crystalline/white


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR