Biometra Thermal Cyclers, Analytik Jena
Supplier: Analytik Jena CA
846-4-070-301 846-2-070-311 846-2-070-301 846-4-070-311 846-4-070-201 846-4-070-251 846-4-070-241 846-4-070-210 846-070-224 846-4-070-280 846-4-070-720 846-4-070-723 846-070-221 846-070-271 846-070-261 846-070-230 846-070-220 846-070-234 846-4-070-212 846-4-070-202 846-4-070-213 846-4-070-215 846-070-232 846-070-222 846-070-233 846-070-235 846-4-070-211 846-4-070-200 846-4-070-224 846-4-070-214 846-070-231
CA75875-684EA 16269.66 CAD
CA75875-684 CA75875-686 CA75875-688 CA75875-864 CA76102-404 CA76102-406 CA76102-408 CA76102-410 CA76102-412 CA76102-414 CA76102-416 CA76102-418 CA76102-452 CA76102-454 CA76102-456 CA76102-458 CA76102-460 CA76102-462 CA76102-464 CA76102-466 CA76102-468 CA76102-470 CA76102-472 CA76102-474 CA76102-476 CA76102-478 CA76102-480 CA76102-548 CA76102-550 CA76102-552 CA76102-554 CA76102-556
Biometra Thermal Cyclers, Analytik Jena
Thermal Cyclers
The Biometra Thermal Cycler product family offers PCR devices ranging from standard to premium equipment – all designed in line with the common principles of being reliable, easy to use, and precise.
  • One block, two block, or 3 block systems available
  • Integrated 7" touchscreen control
  • Whisper quiet, with a low noise emission of 45 dB (max.)
  • High performance smart lid; Defined pressure control for highly reproducible results
  • Linear gradient tool allows easy gradient programming using the primer annealing temperature
  • Heated/Pressurized lid
  • Advanced user
More Product Information