SiliaFlash® Irregular Silica Gels, SiliCycle Inc
Supplier: SiliCycle
R10130D-1KG R10023B-1KG R10030B-1KG R10040B-1KG R10050B-1KG R10170H-1KG R10170M-1KG R10180B-1KG R10180D-1KG R10180H-1KG R12030B-1KG R10150H-1KG R10130H-1KG R10130M-1KG R10137L-1KG R10140A-1KG R10140B-1KG R10140D-1KG R10140H-1KG R10140M-1KG R10160B-1KG R10160H-1KG R10170B-1KG R10170D-1KG R10030B-10KG R10030B-25KG R10140B-5KG R10140B-10KG R10140B-25KG R12030B-5KG R12030B-10KG R12030B-25KG R10030B-5KG R12030B-32KG R10130D-1KG R10023B-1KG R10030B-1KG R10040B-1KG R10050B-1KG R10170H-1KG R10170M-1KG R10180B-1KG R10180D-1KG R10180H-1KG R12030B-1KG R10150H-1KG R10130H-1KG R10130M-1KG R10137L-1KG R10140A-1KG R10140B-1KG R10140D-1KG R10140H-1KG R10140M-1KG R10160B-1KG R10160H-1KG R10170B-1KG R10170D-1KG R10140B-5KG R10140B-10KG R10140B-25KG
75873-682EA 347.03 CAD
75873-682 75873-684 75873-696 75873-698 75873-700 75873-702 75873-704 75873-706 75873-712 75873-714 75873-716 75873-718 75873-720 75873-722 75873-726 75873-728 75873-730 75873-732 75873-734 75873-736 75873-738 75873-740 75873-742 75873-744 75873-746 75873-748 75873-750 75873-752 75873-754 75873-756 76049-248 76049-250 76049-270 76049-284 76049-286 76049-288 76049-290 76049-292 76049-294 76122-202 CA75873-682 CA75873-684 CA75873-696 CA75873-698 CA75873-700 CA75873-702 CA75873-704 CA75873-706 CA75873-712 CA75873-714 CA75873-716 CA75873-718 CA75873-720 CA75873-722 CA75873-726 CA75873-728 CA75873-730 CA75873-732 CA75873-734 CA75873-736 CA75873-738 CA75873-740 CA75873-742 CA75873-744 CA75873-746 CA75873-748 CA75873-750 CA75873-752 CA75873-754 CA75873-756 CA76049-270 CA76049-284 CA76049-286
SiliaFlash® Irregular Silica Gels, SiliCycle Inc
Chromatography Sorbents Flash Chromatography Sorbents
SiliaFlash® products are ideal for both analytical and preparative chromatography, from laboratory to pilot-plant processes and production scales.
  • High purity silica gels
  • Lowest level of fines on the market
  • Exceptional narrow particle and pore size distributions
  • Batch-to-batch, year-to-year consistency
  • Neutral pH
  • Low metal content and controlled water content
With pore diameters ranging from 30 to 1,000 Ångström (Å) and particle sizes up to 1,000 microns (μm), SiliCycle offers products to meet all your requirements. We offer one of the most reliable portfolios for flash and gravity grades for low to medium-high pressure. Our silica gels are ideal for preparative chromatography, from laboratory to pilot-plant processes and production scale.