PURELAB® Chorus 3 Water Purification System, ELGA LabWater
Supplier: ELGA CA
SK-PC2RO310BPIIK SK-PC2RO320BPIIK SK-PC2RO330BPIIK RO330BPM3 SK-CSPPC3-A
CA10034-548EA 8409.82 CAD
CA10034-548 CA76048-800 CA76048-802 CA76048-812 76516-456
PURELAB® Chorus 3 Water Purification System, ELGA LabWater
Water Purification Systems Pure and Ultra Pure Water Systems
When general laboratory grade water (Type III) is all you need, then PURELAB Chorus 3 is the reliable solution with the flexibility to suit your requirements.
  • Ability to configure multiple systems to increase flow rate
  • Simple to install, operate and maintain with a clear indication of water purity
  • Maintains purity of water during periods of low use
  • Optional CO₂ removal from the purified water (post RO) increasing the life of downstream consumables
  • Multiple PURELAB Chorus 3 units can feed into one reservoir and systems can be expanded post installation
  • Cost
More Product Information