Brilliant green solution 1% (w/v) counterstain

Supplier: HARDY DIAGNOSTICS CA

Synonyms: N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulfate, Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Brilliant Green hydrogen sulfate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate

BG008
CA76047-194EA 45.79 CAD
CA76047-194
Brilliant green solution 1% (w/v) counterstain
Brilliant Green

For acid-fast stain procedures and other uses


1% weight/volume. A saline solution of brilliant green.

Formula: C₂₇H₃₄N₂O₄S
MW: 482.65 g/mol
MDL Number: MFCD00011880
CAS Number: 633-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR