Lysing Matrix B, MP Biomedicals
Supplier: MP Biomedicals
116911050 116911100 116911500 116971050 116971100 116971025 116981001 116981010 116911100 116911500 116540425 116540429
75784-614EA 253.7 CAD
75784-614 75784-616 75784-618 CA76045-728 CA76045-730 CA76045-732 CA76045-770 CA76045-772 CA76046-064 CA76046-066 CA76046-224 CA76046-226
Lysing Matrix B, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing Matrix B tubes are used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores
  • Impact resistant tube containing 0.1mm silica spheres
  • Can be used as an additive for disruption by sonication
  • Use for gram-positive and gram-negative bacteria
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then preparedMore Product Information