CoolPrep™ Adapter, MP Biomedicals, LLC
Supplier: MP Biomedicals
116002528
CA76046-178EA 9298.88 CAD
CA76046-178
CoolPrep™ Adapter, MP Biomedicals, LLC
Homogenizers Benchtop Homogenizers
CoolPrep™ Adapter is ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.
  • Consistent, reproducible results
  • For use with FastPrep-24 or FastPrep-96 Homogenizer
  • Ensure efficient cooling of samples
CoolPrep™ 24×2 ml Adapter is a cryogenic adapter which allows simultaneous lysis of 24×2 ml samples. Based on passive temperature control technology, the CoolPrep™ adapter ensures an efficient cooling of the samples as dry-More Product Information