Lysing Matrix D, MP Biomedicals
Supplier: MP Biomedicals
116913100 116913500 116913050 116973025 116973050 116983001 116983010 116913500 116913050 116933050
IC116913100EA 592.55 CAD
IC116913100 75784-606 75784-626 CA76045-734 CA76045-736 CA76045-774 CA76045-776 CA76046-058 CA76046-072 CA76046-190
Lysing Matrix D, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing matrix D is for rapid and reliable biological sample lysis from a wide variety of starting materials.
  • Impact resistant tube containing 1.4mm ceramic spheres
  • Primarily used for the lysis of softer tissues
  • Will also lyse plant tissue, cultured cells and whole insects
  • Grinding matrix is chemically inert and will not bind nucleic acids
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitativeMore Product Information