PHCbi TwinGuard® Series –86 °C Ultra-Low Temperature Upright Freezers, PHC Corporation
Supplier: PHC Corporation
MDF-DU502VXC-PA MDF-DU702VXC-PA MDF-DU302VX-PA MDF-DU502VXC-PA MDF-DU702VXC-PA MDF-DU302VX-PA
76019-346EA 44365.45 CAD
76019-346 76019-348 76200-148 CA76019-346 CA76019-348 CA76200-148
PHCbi TwinGuard® Series –86 °C Ultra-Low Temperature Upright Freezers, PHC Corporation
Freezers Ultra Low Temperature Freezers
The PHCbi Twin Guard® Series satisfies an industry demand for safe, long-term storage for the most high-valued materials.
  • Two refrigeration systems
  • Ideal for storing sensitive specimens
  • EcoMode functionality
  • Five-year manufacturer's parts and labor warranty
Two independent refrigeration systems, combined with an optional liquid CO₂ back-up system, offer a circle of protection unmatched in the marketplace
The two TwinGuard® Series' compressors are completely separated from one another with no reliance on one another to operate. In the unlikely eventMore Product Information