PLET Agar, for Bacillus anthracis
Supplier: HARDY DIAGNOSTICS CA
G153
CA76006-290PK 190.88 CAD
CA76006-290
PLET Agar, for Bacillus anthracis
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of Bacillus anthracis from environmental samples, animal products, carcasses, and clinical samples from non-sterile sites.
Order Now
SPECIFICATIONS
Environmentally Preferable