Bottle Top Dispensers, Sartorius
Supplier: Sartorius
LH-723071 LH-723072 LH-723073 LH-723074 LH-723075 LH-723060 LH-723061 LH-723062 LH-723063 LH-723064 LH-723065 LH-723070
CA75994-986EA 1091.68 CAD
CA75994-986 CA75994-988 CA75994-990 CA75994-992 CA75994-994 CA75995-126 CA75995-128 CA75995-130 CA75995-132 CA75995-134 CA75995-136 CA75995-138
Bottle Top Dispensers, Sartorius
Bottle Top Dispensers
Prospenser and Prospenser Plus are high quality bottle-top dispensers that offer excellent chemical resistance.
  • Incremental volume setup ensures accurate and repeatable results
  • Fully autoclavable
  • Smooth and light to operate
  • Easy to attach aspiration tube with a secure threaded connection
  • User-replaceable valves and wide range of accessories are available
Adavanced features of Prospenser Plus include, a media recirculation system that allows unused media to be recovered from the device and returned to the bottle, 360° rotating dispensing head, and aMore Product Information