Biotrate Digital Burettes
Supplier: Sartorius
LH-723080 LH-723081 LH-723082
CA75994-996EA 2477.84 CAD
CA75994-996 CA75994-998 CA75995-000
Biotrate Digital Burettes
Burettes
Biotrate is a premium digital burette equipped with sophisticated functions for easy and convenient titration. Biotrate offers high chemical resistance and it is a smart solution for titrating various liquids accurately and safely.
  • Manual titration of liquids
  • Extremely high chemical resistance
  • Smooth and light to operate with soft touch operating wheels
  • 360° rotating dispensing head
  • Universal connector at the back of the unit for drying tubes and filters
  • User-replaceable valves
  • Long life CR2032 battery
  • Wide range of accessories
Media recirculationMore Product Information