SiliaBond® DMAP, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R75630B-100G R75630B-10G R75630B-1KG R75630B-250G R75630B-25G R75630B-500G R75630B-50G R75630B-100G R75630B-10G R75630B-1KG R75630B-250G R75630B-25G R75630B-500G R75630B-50G
75991-058EA 3420.56 CAD
75991-058 75991-060 75991-062 75991-064 75991-066 75991-068 75991-070 75991-072 CA75991-058 CA75991-060 CA75991-062 CA75991-064 CA75991-066 CA75991-068 CA75991-070 CA75991-072
SiliaBond® DMAP, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.
Order Now
SPECIFICATIONS
Environmentally Preferable