SiliaBond® C18, Polymeric, Functionalized Irregular Silica Gel, 17%, SiliCycle
Supplier: Avantor
R00230B-500G R00230B-1KG R00230B-250G R00230B-50G R00230B-10G R00230B-1KG
75990-586EA 587.44 CAD
75990-586 75990-588 75990-590 75990-592 75990-594 75990-596 75991-778 75991-780 CA75990-586 CA75990-588 CA75990-590 CA75990-592 CA75990-594 CA75990-596 CA75991-778 CA75991-780
SiliaBond® C18, Polymeric, Functionalized Irregular Silica Gel, 17%, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.