BioClean-D™ Sterile Extra-Length Overboots, Ansell
Supplier: Ansell Canada
BDOB S-BDOB BDOB S-BDOB BDOB-L
75972-644CS 932.44 CAD
75972-644 75982-216 CA75972-644 CA75982-216 77474-000
BioClean-D™ Sterile Extra-Length Overboots, Ansell
Disposable Garments
These disposable boot covers are made of BioClean-D™, a lightweight, low linting and hard-wearing fabric (PP laminated with a PE film) which provides maximum comfort and protects against fine sprays and particles.
  • Longer length: 500 mm
  • Low-linting
  • Tie-fastenings at top and ankle
  • Slip-resistant polyurethane sole
  • Latex-free
  • Available as sterile or nonsterile version
  • Elasticated top with secure ties
  • A unique barcode on the outer carton (ITF-14) allows automatic data entry into Stock Control Systems
Overboots feature a slip resistant sole and easy tie-More Product Information