BioClean Fusion™ Cleanroom Gloves, Polychloroprene, Ansell
Supplier: Ansell Canada
BFAP-L BFAP-XS BFAP-XXL BFAP-M BFAP-S BFAP-XL S-BFAP-5055 S-BFAP-6065 S-BFAP-7075 S-BFAP-8085 S-BFAP-9090 BFAP-L BFAP-XS BFAP-XXL BFAP-M BFAP-S BFAP-XL S-BFAP-1010 S-BFAP-5055 S-BFAP-6065 S-BFAP-7075 S-BFAP-8085 S-BFAP-9090
75972-612CS 1027.06 CAD
75972-612 75972-620 75972-622 75972-650 75972-652 75972-654 75982-296 75982-304 75982-306 75982-308 75982-310 75982-312 CA75972-612 CA75972-620 CA75972-622 CA75972-650 CA75972-652 CA75972-654 CA75982-296 CA75982-304 CA75982-306 CA75982-308 CA75982-310 CA75982-312
BioClean Fusion™ Cleanroom Gloves, Polychloroprene, Ansell
Gloves
BioClean Fusion™ polychloroprene cleanroom gloves are comfortable and thin to reduce wearer fatigue and increase tactility.
  • Sterility assurance level 10⁻⁶
  • The anti-static glove is double donnable
  • Increased tactility
  • Beaded cuff for strength
  • Non-particulating EasyTear™ packaging
External glove surface for textured fingers with chlorinated internal glove surface.
Certifications: Manufactured in a facility holding ISO 9001: 2008 and 89/686/EEC article 11b certifications
Conforms to category 3 - complex design personal protective equipment PPE directiveMore Product Information