SiliaSep™ Imidazole Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR79230BI1500 FR79230BIS120 FR79230BIS220 FR79230BIS330 FR79230BIS750 FR79230BISO04 FR79230BISO12 FR79230BISO25 FR79230BISO40 FR79230BISO80 FR79230BI1500 FR79230BIS120 FR79230BIS220 FR79230BIS330 FR79230BIS750 FR79230BISO04 FR79230BISO12 FR79230BISO25 FR79230BISO40 FR79230BISO80
75880-274BX 21443.79 CAD
75880-274 75880-276 75880-278 75880-280 75880-282 75880-284 75880-286 75880-288 75880-290 75880-292 CA75880-274 CA75880-276 CA75880-278 CA75880-280 CA75880-282 CA75880-284 CA75880-286 CA75880-288 CA75880-290 CA75880-292
SiliaSep™ Imidazole Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offersMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable