SiliaSep™ Triamine Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR48030BIS750 FR48030BISO04 FR48030BISO12 FR48030BISO25 FR48030BISO40 FR48030BISO80 FR48030BI1500 FR48030BIS120 FR48030BIS220 FR48030BIS330 FR48030BIS750 FR48030BISO04 FR48030BISO12 FR48030BISO25 FR48030BISO40 FR48030BISO80 FR48030BI1500 FR48030BIS120 FR48030BIS220 FR48030BIS330
75879-362BX 7442.34 CAD
75879-362 75879-364 75879-366 75879-368 75879-370 75879-372 75879-970 75879-972 75879-974 75879-976 CA75879-362 CA75879-364 CA75879-366 CA75879-368 CA75879-370 CA75879-372 CA75879-970 CA75879-972 CA75879-974 CA75879-976
SiliaSep™ Triamine Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offersMore Product Information