Brady® BMP®61 Series Printer Ribbon
Supplier: Brady Worldwide
148128
CA75840-073RL 45.45 CAD
CA75840-073
Brady® BMP®61 Series Printer Ribbon
Printer Ribbons
BMP®61 Series ribbon cartridges allow users to change ribbons in seconds.
  • R4300 Series Ribbons offers strong environmental resistance against smearing and chemicals
  • R6000 Series Halogen-Free Resin formulation offers excellent solvent, scratch, and heat resistance
  • R6200 Series Ribbons offer superior scratch, smear, heat, and chemical resistance
  • R6400 Series Resin formulation offers superior chemical and abrasion resistance
  • R6600 Series Resin formulation offers excellent solvent and friction resistance
  • R6700 Series Resin/Wax formulation is a white ribbon
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable